siRNA Details
Gene Symbol: ERI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142725.1 | NP_001136197.1 | 6 | 537 | ||
NM_080663.2 | NP_542394.2 | 6 | 537 | |||
GenBank mRNA | AK127391.1 | - | 6 | 540 | ||
AK292594.1 | - | 7 | 750 | |||
BX437401.2 | - | 3 | 226 |

More Information
Other Species Targeted

Gene Ontology Categories:
