siRNA Details
Gene Symbol: ZNF512B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020713.2 | NP_065764.1 | 17 | 5253 | ||
XM_005260226.3 | XP_005260283.1 | 18 | 5325 | |||
XM_011528929.2 | XP_011527231.1 | 17 | 5215 | |||
XM_011528930.2 | XP_011527232.1 | 16 | 5343 | |||
XM_011528931.2 | XP_011527233.1 | 13 | 4658 | |||
XM_011528932.2 | XP_011527234.1 | 13 | 4801 | |||
GenBank mRNA | AB033022.1 | - | 15 | 5074 | ||
AL834525.1 | - | 11 | 3923 |
Gene Symbol: UCKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011528869.1 | XP_011527171.1 | 1 | 2063 | ||
XM_011528872.1 | XP_011527174.1 | 1 | 2063 | |||
XM_017027904.1 | XP_016883393.1 | 1 | 2063 | |||
XM_017027905.1 | XP_016883394.1 | 1 | 2063 | |||
XM_017027906.1 | XP_016883395.1 | 1 | 2063 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
