siRNA Details
Gene Symbol: ZGPAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083113.1 | NP_001076582.1 | 7 | 1745 | ||
NM_001195653.1 | NP_001182582.1 | 7 | 1769 | |||
NM_001195654.1 | NP_001182583.1 | 7 | 1761 | |||
NM_032527.4 | NP_115916.3 | 7 | 1848 | |||
NM_181485.2 | NP_852150.2 | 7 | 1718 | |||
GenBank mRNA | AB058750.3 | - | 7 | 1722 | ||
AK027878.1 | - | 7 | 1786 | |||
AK074961.1 | - | 6 | 2672 | |||
AK307368.1 | - | 3 | 653 | |||
BC019338.1 | - | 7 | 1736 | 2906985 | ||
BC032612.1 | - | 7 | 1708 | 5555076 |
Gene Symbol: LIME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305654.1 | NP_001292583.1 | 1 | 282 | ||
GenBank mRNA | AY203957.1 | - | 1 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
