siRNA Details
Gene Symbol: ZGPAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083113.1 | NP_001076582.1 | 6 | 1486 | ||
NM_001195653.1 | NP_001182582.1 | 6 | 1510 | |||
NM_001195654.1 | NP_001182583.1 | 6 | 1502 | |||
NM_032527.4 | NP_115916.3 | 6 | 1589 | |||
NM_181485.2 | NP_852150.2 | 6 | 1459 | |||
GenBank mRNA | AB058750.3 | - | 6 | 1463 | ||
AK027878.1 | - | 6 | 1527 | |||
AK074961.1 | - | 5 | 2413 | |||
AK307368.1 | - | 2 | 394 | |||
AK314726.1 | - | 6 | 1499 | |||
BC019338.1 | - | 6 | 1477 | 2906985 | ||
BC032612.1 | - | 6 | 1449 | 5555076 | ||
KU178806.1 | - | 5 | 1340 | |||
KU178807.1 | - | 5 | 1376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
