siRNA Details
Gene Symbol: ZGPAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083113.1 | NP_001076582.1 | 2 | 455 | ||
NM_001195653.1 | NP_001182582.1 | 2 | 479 | |||
NM_001195654.1 | NP_001182583.1 | 2 | 498 | |||
NM_032527.4 | NP_115916.3 | 2 | 498 | |||
NM_181485.2 | NP_852150.2 | 2 | 428 | |||
GenBank mRNA | AB058750.3 | - | 2 | 432 | ||
AK027878.1 | - | 2 | 436 | |||
AK074961.1 | - | 2 | 417 | |||
AK314726.1 | - | 2 | 468 | |||
BC019338.1 | - | 2 | 473 | 2906985 | ||
BC032612.1 | - | 2 | 418 | 5555076 | ||
CV027894.1 | - | 1 | 309 | |||
DA055150.1 | - | 2 | 479 | |||
DA252406.1 | - | 2 | 455 | |||
DA848516.1 | - | 2 | 429 | |||
DB025637.1 | - | 2 | 487 | |||
KU178806.1 | - | 1 | 309 | |||
KU178807.1 | - | 1 | 309 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
