siRNA Details
Gene Symbol: ZNF655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009960.1 | NP_001009960.1 | 3 | 1924 | ||
NM_001083956.1 | NP_001077425.1 | 4 | 2202 | |||
NM_001085368.1 | NP_001078837.1 | 4 | 2029 | |||
NM_138494.2 | NP_612503.1 | 3 | 2097 | |||
XM_017012598.1 | XP_016868087.1 | 6 | 3102 | |||
XM_017012599.1 | XP_016868088.1 | 6 | 2260 | |||
XM_017012600.1 | XP_016868089.1 | 6 | 2273 | |||
XM_017012601.1 | XP_016868090.1 | 6 | 2259 | |||
XM_017012602.1 | XP_016868091.1 | 5 | 2997 | |||
XM_017012603.1 | XP_016868092.1 | 4 | 3122 | |||
XM_017012604.1 | XP_016868093.1 | 4 | 2039 | |||
XM_017012605.1 | XP_016868094.1 | 5 | 2050 | |||
XM_017012606.1 | XP_016868095.1 | 3 | 1955 | |||
XM_017012607.1 | XP_016868096.1 | 3 | 1941 | |||
XM_017012608.1 | XP_016868097.1 | 6 | 3135 | |||
XM_017012609.1 | XP_016868098.1 | 5 | 2149 | |||
XM_017012610.1 | XP_016868099.1 | 5 | 2096 | |||
GenBank mRNA | AK122929.1 | - | 1 | 1181 | ||
AK128057.1 | - | 2 | 3161 | |||
AK316024.1 | - | 4 | 2020 | |||
AY099353.1 | - | 3 | 1923 | |||
BC011816.1 | - | 1 | 1192 | |||
BC024770.1 | - | 1 | 1443 | |||
BC037407.1 | - | 3 | 1887 | 5752559 | ||
CR749270.1 | - | 3 | 1705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
