siRNA Details
Gene Symbol: POLR2J3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097615.2 | NP_001091084.2 | 8 | 1015 | ||
XM_017012373.1 | XP_016867862.1 | 9 | 1209 |
Gene Symbol: LOC107983993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012895.1 | XP_016868384.1 | 6 | 762 | ||
XM_017012896.1 | XP_016868385.1 | 6 | 1285 |
Gene Symbol: POLR2J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032959.5 | NP_116581.3 | 8 | 1043 | ||
GenBank mRNA | BC056864.1 | - | 8 | 933 | ||
BC086857.1 | - | 7 | 868 | 6007730 |
Gene Symbol: UPK3BL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114403.2 | NP_001107875.1 | 5 | 675 | ||
GenBank mRNA | EU341824.1 | - | 5 | 675 | ||
KF150200.1 | - | 5 | 642 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
