siRNA Details
Gene Symbol: SSPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198455.2 | NP_940857.2 | 16 | 2381 | ||
GenBank mRNA | AK127639.1 | - | 5 | 676 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
