siRNA Details
Gene Symbol: CFAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025145.5 | NP_079421.5 | 32 | 4248 | ||
XM_005270171.2 | XP_005270228.1 | 32 | 4252 | |||
XM_005270172.3 | XP_005270229.1 | 31 | 4165 | |||
XM_011540196.2 | XP_011538498.1 | 33 | 4366 | |||
XM_011540197.2 | XP_011538499.1 | 32 | 4279 | |||
XM_011540198.2 | XP_011538500.1 | 32 | 4249 | |||
XM_011540199.2 | XP_011538501.1 | 32 | 4249 | |||
XM_011540201.2 | XP_011538503.1 | 33 | 4366 | |||
XM_011540202.2 | XP_011538504.1 | 29 | 3682 | |||
XM_011540203.2 | XP_011538505.1 | 31 | 3924 | |||
XM_017016681.1 | XP_016872170.1 | 33 | 4363 | |||
XM_017016682.1 | XP_016872171.1 | 30 | 4018 | |||
XM_017016683.1 | XP_016872172.1 | 30 | 3804 | |||
GenBank mRNA | AK093325.1 | - | 6 | 678 | ||
AL833836.1 | - | 17 | 2127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
