siRNA Details
Gene Symbol: CFAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025145.5 | NP_079421.5 | 18 | 2459 | ||
XM_005270171.2 | XP_005270228.1 | 18 | 2463 | |||
XM_005270172.3 | XP_005270229.1 | 18 | 2463 | |||
XM_011540196.2 | XP_011538498.1 | 19 | 2577 | |||
XM_011540197.2 | XP_011538499.1 | 19 | 2577 | |||
XM_011540198.2 | XP_011538500.1 | 18 | 2460 | |||
XM_011540199.2 | XP_011538501.1 | 19 | 2577 | |||
XM_011540200.2 | XP_011538502.1 | 19 | 2577 | |||
XM_011540201.2 | XP_011538503.1 | 19 | 2577 | |||
XM_011540202.2 | XP_011538504.1 | 15 | 1893 | |||
XM_011540203.2 | XP_011538505.1 | 17 | 2135 | |||
XM_017016681.1 | XP_016872170.1 | 19 | 2574 | |||
XM_017016682.1 | XP_016872171.1 | 17 | 2346 | |||
XM_017016683.1 | XP_016872172.1 | 16 | 2015 | |||
XM_017016684.1 | XP_016872173.1 | 18 | 2463 | |||
GenBank mRNA | AK295018.1 | - | 18 | 2441 | ||
AL833836.1 | - | 4 | 425 | |||
BC036647.2 | - | 18 | 2462 | 4825754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
