siRNA Details
Gene Symbol: ZNF44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164276.1 | NP_001157748.1 | 5 | 2075 | ||
NM_016264.3 | NP_057348.3 | 4 | 1931 | |||
XM_005259934.3 | XP_005259991.1 | 4 | 1941 | |||
XM_011528062.2 | XP_011526364.1 | 5 | 2040 | |||
XM_011528064.1 | XP_011526366.1 | 4 | 2356 | |||
XM_011528066.1 | XP_011526368.1 | 3 | 2229 | |||
XM_017026870.1 | XP_016882359.1 | 4 | 2035 | |||
XM_017026871.1 | XP_016882360.1 | 5 | 2074 | |||
GenBank mRNA | AB021642.1 | - | 4 | 1866 | ||
AK297528.1 | - | 5 | 1989 | |||
BC136502.1 | - | 5 | 1970 | 9020491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
