siRNA Details
Gene Symbol: PCNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014801.3 | NP_055616.3 | 13 | 3082 | ||
XM_006711816.3 | XP_006711879.1 | 13 | 2904 | |||
XM_011544278.2 | XP_011542580.2 | 6 | 693 | |||
XM_017002391.1 | XP_016857880.1 | 12 | 2646 | |||
XM_017002392.1 | XP_016857881.1 | 1 | 83 | |||
GenBank mRNA | AK021445.1 | - | 7 | 1058 | ||
AK074078.1 | - | 8 | 2187 | |||
AK092353.1 | - | 7 | 1478 | |||
BC008300.1 | - | 8 | 992 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
