siRNA Details
Gene Symbol: MBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025081.1 | NP_001020252.1 | 7 | 2003 | ||
NM_001025090.1 | NP_001020261.1 | 6 | 1925 | |||
NM_001025092.1 | NP_001020263.1 | 5 | 1892 | |||
NM_001025101.1 | NP_001020272.1 | 9 | 2497 | |||
NM_002385.2 | NP_002376.1 | 6 | 1970 | |||
GenBank mRNA | AK094611.1 | - | 1 | 2089 | ||
AK095121.1 | - | 1 | 2881 | |||
AK098402.1 | - | 6 | 1881 | |||
AK124830.1 | - | 2 | 3222 | |||
AK126858.1 | - | 1 | 5035 | |||
AK128770.1 | - | 6 | 1884 | |||
AK128788.1 | - | 6 | 1881 | |||
BC008749.2 | - | 6 | 1863 | 3346521 | ||
BC068550.1 | - | 4 | 1739 | |||
BC080654.1 | - | 5 | 1839 | 4859683 | ||
CR627018.1 | - | 4 | 9651 | |||
M13577.1 | - | 6 | 1870 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
