siRNA Details
Gene Symbol: MBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025100.1 | NP_001020271.1 | 2, 3 | 326 | ||
NM_001025101.1 | NP_001020272.1 | 2, 3 | 326 | |||
XM_011526009.2 | XP_011524311.1 | 2, 3 | 280 | |||
XM_017025779.1 | XP_016881268.1 | 2, 3 | 1005 | |||
XM_017025780.1 | XP_016881269.1 | 2, 3 | 280 | |||
GenBank mRNA | AB208986.1 | - | 2, 3 | 277 | ||
BC065248.1 | - | 2, 3 | 266 | 6060520 | ||
L18865.1 | - | 2, 3 | 277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
