siRNA Details
Gene Symbol: MBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025081.1 | NP_001020252.1 | 7 | 725 | ||
NM_001025090.1 | NP_001020261.1 | 6 | 647 | |||
NM_001025092.1 | NP_001020263.1 | 5 | 614 | |||
NM_001025101.1 | NP_001020272.1 | 9 | 1219 | |||
NM_002385.2 | NP_002376.1 | 6 | 692 | |||
GenBank mRNA | AK094611.1 | - | 1 | 811 | ||
AK095121.1 | - | 1 | 1601 | |||
AK098402.1 | - | 6 | 603 | |||
AK123433.1 | - | 2 | 3267 | |||
AK124830.1 | - | 2 | 1944 | |||
AK126858.1 | - | 1 | 3757 | |||
AK128770.1 | - | 6 | 606 | |||
AK128788.1 | - | 6 | 603 | |||
AK289893.1 | - | 6 | 647 | |||
AK293922.1 | - | 5 | 642 | |||
BC008749.2 | - | 6 | 585 | 3346521 | ||
CR627018.1 | - | 4 | 8373 | |||
M13577.1 | - | 6 | 592 | |||
M30515.1 | - | 8 | 641 | |||
M30516.1 | - | 6 | 611 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
