siRNA Details
Gene Symbol: MT1H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005951.2 | NP_005942.1 | 1 | 56 | ||
GenBank mRNA | BC008408.1 | - | 1 | 56 | 4274128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
