siRNA Details
Gene Symbol: SYS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099791.2 | NP_001093261.1 | 1 | 197 | ||
NM_001197129.1 | NP_001184058.1 | 2 | 221 | |||
NM_033542.3 | NP_291020.1 | 1 | 197 | |||
GenBank mRNA | AK075264.1 | - | 1 | 160 | ||
AK172738.1 | - | 1 | 164 | |||
AK309429.1 | - | 1 | 67 | |||
AL591714.1 | - | 1 | 92 | |||
BC048286.1 | - | 1 | 127 | 5212523 | ||
BI820996.1 | - | 2 | 221 | |||
DA526637.1 | - | 1 | 197 |
Gene Symbol: SYS1-DBNDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003189.2 | 1 | 128 | |||
GenBank mRNA | BC023562.2 | - | 1 | 97 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
