siRNA Details
Gene Symbol: SYS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197129.1 | NP_001184058.1 | 5 | 1338 | ||
NM_033542.3 | NP_291020.1 | 4 | 1314 | |||
GenBank mRNA | AK075264.1 | - | 4 | 1277 | ||
AL591714.1 | - | 4 | 1209 | |||
BC048286.1 | - | 4 | 1244 | 5212523 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
