siRNA Details
Gene Symbol: APOC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000040.1 | NP_000031.1 | 1 | 15 | ||
GenBank mRNA | BC027977.1 | - | 1 | 13 | 5229117 | |
BC121081.1 | - | 1 | 15 | 40119733 | ||
BC134419.1 | - | 1 | 15 | 40119734 | ||
X01388.1 | - | 1 | 40 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
