siRNA Details
Gene Symbol: GPSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276501.1 | NP_001263430.1 | 1 | 431 | ||
NM_022107.2 | NP_071390.1 | 5 | 402 | |||
GenBank mRNA | AF155657.1 | - | 1 | 50 | ||
AJ243937.1 | - | 5 | 402 | |||
AJ243938.1 | - | 1 | 422 | |||
AK313922.1 | - | 1 | 431 | |||
BC018724.1 | - | 1 | 94 | 4576337 | ||
DA979033.1 | - | 1 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
