siRNA Details
Gene Symbol: PRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005039.3 | NP_005030.2 | 3 | 502 | ||
NM_199353.2 | NP_955385.1 | 4 | 472 | |||
NM_199354.2 | NP_955386.1 | 4 | 412 | |||
GenBank mRNA | BC044827.1 | - | 3 | 488 | ||
BG216715.1 | - | 1 | 185 | |||
BX487087.1 | - | 3 | 501 | |||
K03204.1 | - | 3 | 502 |
Gene Symbol: PRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006248.3 | NP_006239.3 | 3 | 380 | ||
GenBank mRNA | BX484538.1 | - | 3 | 380 | ||
K03208.1 | - | 1 | 36 |
Gene Symbol: PRB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002723.5 | NP_002714.2 | 3 | 577 | ||
GenBank mRNA | BC035362.1 | - | 3 | 626 | ||
BC130386.1 | - | 3 | 625 | 40146417 | ||
K03207.1 | - | 3 | 576 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
