siRNA Details
Gene Symbol: PRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005039.3 | NP_005030.2 | 3 | 262 | ||
NM_199353.2 | NP_955385.1 | 3 | 262 | |||
NM_199354.2 | NP_955386.1 | 3 | 262 | |||
GenBank mRNA | BC044827.1 | - | 3 | 248 | ||
BC141917.1 | - | 3 | 262 | 40113876 | ||
BG216715.1 | - | 1 | 128 | |||
BX487087.1 | - | 3 | 261 | |||
K03204.1 | - | 3 | 262 | |||
K03205.1 | - | 3 | 250 | |||
K03206.1 | - | 3 | 258 |
Gene Symbol: PRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006248.3 | NP_006239.3 | 3 | 323 | ||
GenBank mRNA | BX484538.1 | - | 3 | 323 | ||
K03208.1 | - | 1 | 162 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
