siRNA Details
Gene Symbol: NUDT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304963.1 | NP_001291892.1 | 1 | 358 | ||
NM_153183.3 | NP_694853.1 | 2 | 421 | |||
GenBank mRNA | AF469196.1 | - | 2 | 423 | ||
AK291952.1 | - | 1 | 338 | |||
BC050700.1 | - | 1 | 337 | 5723622 | ||
BX449265.2 | - | 1 | 358 | |||
CN399689.1 | - | 1 | 27 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
