siRNA Details
Gene Symbol: RHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127691.2 | NP_001121163.1 | 7 | 1054 | ||
NM_001282867.1 | NP_001269796.1 | 10 | 1338 | |||
NM_001282868.1 | NP_001269797.1 | 8 | 1134 | |||
NM_001282869.1 | NP_001269798.1 | 8 | 1188 | |||
NM_001282870.1 | NP_001269799.1 | 9 | 1268 | |||
NM_001282871.1 | NP_001269800.1 | 9 | 1358 | |||
NM_001282872.1 | NP_001269801.1 | 10 | 1438 | |||
NM_016124.4 | NP_057208.2 | 10 | 1342 | |||
GenBank mRNA | AB049754.1 | - | 10 | 1280 | ||
AF187846.1 | - | 10 | 1280 | |||
AF312679.1 | - | 10 | 1315 | |||
AF510069.1 | - | 10 | 1323 | |||
AF510070.1 | - | 10 | 1323 | |||
AY751491.1 | - | 7 | 992 | |||
AY751492.1 | - | 8 | 1072 | |||
AY751493.1 | - | 8 | 1126 | |||
AY751494.1 | - | 9 | 1206 | |||
AY751495.1 | - | 9 | 1296 | |||
AY751496.1 | - | 10 | 1376 | |||
BC139922.1 | - | 7 | 1042 | 40148684 | ||
DQ451878.1 | - | 10 | 1299 | |||
FR745438.1 | - | 10 | 1280 | |||
FR745439.1 | - | 10 | 1280 | |||
FR745440.1 | - | 10 | 1280 | |||
FR745441.1 | - | 10 | 1280 | |||
JQ424879.1 | - | 10 | 1338 | |||
L08429.1 | - | 10 | 1280 | |||
LN824132.1 | - | 10 | 1280 | |||
X63094.1 | - | 10 | 1338 | |||
X63097.1 | - | 10 | 1296 | |||
Z97026.1 | - | 10 | 1300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
