siRNA Details
Gene Symbol: RHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127691.2 | NP_001121163.1 | 2, 3 | 393 | ||
NM_001282867.1 | NP_001269796.1 | 2, 3 | 393 | |||
NM_001282868.1 | NP_001269797.1 | 2, 3 | 393 | |||
NM_001282869.1 | NP_001269798.1 | 2, 3 | 393 | |||
NM_001282870.1 | NP_001269799.1 | 2, 3 | 393 | |||
NM_001282871.1 | NP_001269800.1 | 2, 3 | 393 | |||
NM_001282872.1 | NP_001269801.1 | 2, 3 | 393 | |||
NM_016124.4 | NP_057208.2 | 2, 3 | 393 | |||
XM_017002015.1 | XP_016857504.1 | 2, 3 | 486 | |||
GenBank mRNA | AB018966.1 | - | 2, 3 | 331 | ||
AB018967.1 | - | 2, 3 | 331 | |||
AB018968.1 | - | 2, 3 | 331 | |||
AB018969.1 | - | 2, 3 | 331 | |||
AB037270.1 | - | 2, 3 | 331 | |||
AB046420.1 | - | 2, 3 | 331 | |||
AB049754.1 | - | 2, 3 | 331 | |||
AF037626.1 | - | 2, 3 | 331 | |||
AF177938.1 | - | 2, 3 | 331 | |||
AF187846.1 | - | 2, 3 | 331 | |||
AF312679.1 | - | 2, 3 | 366 | |||
AF510069.1 | - | 2, 3 | 374 | |||
AF510070.1 | - | 2, 3 | 374 | |||
AJ704215.2 | - | 2, 3 | 331 | |||
AK297151.1 | - | 2, 3 | 486 | |||
AM183924.3 | - | 2, 3 | 331 | |||
AY449381.1 | - | 2, 3 | 331 | |||
AY449382.1 | - | 2, 3 | 331 | |||
AY449383.1 | - | 2, 3 | 331 | |||
AY449384.1 | - | 2, 3 | 331 | |||
AY449385.1 | - | 2, 3 | 331 | |||
AY751491.1 | - | 2, 3 | 331 | |||
AY751492.1 | - | 2, 3 | 331 | |||
AY751493.1 | - | 2, 3 | 331 | |||
AY751494.1 | - | 2, 3 | 331 | |||
AY751495.1 | - | 2, 3 | 331 | |||
AY751496.1 | - | 2, 3 | 331 | |||
BC139922.1 | - | 2, 3 | 381 | 40148684 | ||
DQ451878.1 | - | 2, 3 | 350 | |||
EU557240.1 | - | 2, 3 | 334 | |||
EU557241.1 | - | 2, 3 | 334 | |||
EU661905.1 | - | 2, 3 | 271 | |||
EU661906.1 | - | 2, 3 | 271 | |||
FR745438.1 | - | 2, 3 | 331 | |||
FR745439.1 | - | 2, 3 | 331 | |||
FR745440.1 | - | 2, 3 | 331 | |||
FR745441.1 | - | 2, 3 | 331 | |||
HE657775.1 | - | 2, 3 | 331 | |||
JF436967.1 | - | 2, 3 | 331 | |||
JN635688.1 | - | 2, 3 | 331 | |||
JN635689.1 | - | 2, 3 | 331 | |||
JQ065733.1 | - | 2, 3 | 331 | |||
JQ424879.1 | - | 2, 3 | 389 | |||
KT240043.1 | - | 2, 3 | 331 | |||
L08429.1 | - | 2, 3 | 331 | |||
LC053447.1 | - | 2, 3 | 331 | |||
LN714468.1 | - | 2, 3 | 331 | |||
LN824132.1 | - | 2, 3 | 331 | |||
S57971.1 | - | 2, 3 | 331 | |||
S70174.1 | - | 2, 3 | 331 | |||
S73913.1 | - | 2, 3 | 331 | |||
S78509.1 | - | 2, 3 | 331 | |||
S82449.1 | - | 2, 3 | 331 | |||
X63094.1 | - | 2, 3 | 389 | |||
X63097.1 | - | 2, 3 | 347 | |||
Z97026.1 | - | 2, 3 | 351 |
Gene Symbol: RHCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020485.4 | NP_065231.3 | 2, 3 | 417 | ||
NM_138616.3 | NP_619522.3 | 2, 3 | 417 | |||
NM_138617.3 | NP_619523.3 | 2, 3 | 417 | |||
NM_138618.3 | NP_619524.3 | 2, 3 | 417 | |||
XM_005245957.3 | XP_005246014.1 | 2, 3 | 882 | |||
XM_006710810.3 | XP_006710873.1 | 2, 3 | 886 | |||
XM_011541888.2 | XP_011540190.1 | 3, 4 | 917 | |||
XM_011541889.2 | XP_011540191.1 | 2, 3 | 432 | |||
XM_011541890.2 | XP_011540192.1 | 2, 3 | 878 | |||
XM_011541891.2 | XP_011540193.1 | 3, 4 | 534 | |||
XM_017002014.1 | XP_016857503.1 | 2, 3 | 885 | |||
GenBank mRNA | AB018644.1 | - | 2, 3 | 331 | ||
AB018645.1 | - | 2, 3 | 331 | |||
AB030388.1 | - | 2, 3 | 331 | |||
AB049753.1 | - | 2, 3 | 331 | |||
AF056965.1 | - | 2, 3 | 331 | |||
AF510065.1 | - | 2, 3 | 374 | |||
AF510066.1 | - | 2, 3 | 374 | |||
AF510068.1 | - | 2, 3 | 374 | |||
AM295506.2 | - | 2, 3 | 331 | |||
AY603478.1 | - | 2, 3 | 347 | |||
BC075081.2 | - | 2, 3 | 352 | 30915384 | ||
BC139905.1 | - | 2, 3 | 459 | 40147407 | ||
DQ178642.1 | - | 2, 3 | 331 | |||
DQ266353.1 | - | 2, 3 | 331 | |||
DQ266400.1 | - | 2, 3 | 331 | |||
DQ322275.1 | - | 2, 3 | 331 | |||
GU474431.1 | - | 2, 3 | 331 | |||
GU563377.1 | - | 2, 3 | 331 | |||
GU810838.1 | - | 2, 3 | 331 | |||
HE572747.1 | - | 2, 3 | 331 | |||
HG326207.1 | - | 2, 3 | 331 | |||
HQ711843.1 | - | 2, 3 | 331 | |||
HQ711844.1 | - | 2, 3 | 351 | |||
HQ711845.1 | - | 2, 3 | 331 | |||
HQ711846.1 | - | 2, 3 | 331 | |||
HQ711847.1 | - | 2, 3 | 351 | |||
JF436968.1 | - | 2, 3 | 331 | |||
JX420291.1 | - | 2, 3 | 331 | |||
JX420292.1 | - | 2, 3 | 331 | |||
JX420293.1 | - | 2, 3 | 331 | |||
JX420294.1 | - | 2, 3 | 331 | |||
JX420295.1 | - | 2, 3 | 331 | |||
JX420296.1 | - | 2, 3 | 331 | |||
JX420297.1 | - | 2, 3 | 331 | |||
JX420298.1 | - | 2, 3 | 331 | |||
JX420299.1 | - | 2, 3 | 331 | |||
JX420300.1 | - | 2, 3 | 331 | |||
JX420302.1 | - | 2, 3 | 331 | |||
KF515558.1 | - | 2, 3 | 331 | |||
KM975480.1 | - | 2, 3 | 331 | |||
KP056313.1 | - | 2, 3 | 331 | |||
M34015.1 | - | 2, 3 | 361 | |||
S57967.1 | - | 2, 3 | 331 | |||
X54534.1 | - | 2, 3 | 375 | |||
X63095.1 | - | 2, 3 | 340 | |||
X63096.1 | - | 2, 3 | 337 | |||
X63098.1 | - | 2, 3 | 350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
