siRNA Details
Gene Symbol: OR11H12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013354.1 | NP_001013372.1 | 1 | 315 | ||
GenBank mRNA | BC136730.1 | - | 1 | 387 | 9020720 | |
BC136731.1 | - | 1 | 387 | 9020721 |
Gene Symbol: OR11H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005239.1 | NP_001005239.1 | 1 | 315 |
Gene Symbol: OR11H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197287.1 | NP_001184216.1 | 2 | 387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
