siRNA Details
Gene Symbol: ERGIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031711.2 | NP_001026881.1 | 5 | 466 | ||
XM_006714892.1 | XP_006714955.1 | 3 | 157 | |||
XM_011534597.1 | XP_011532899.1 | 5 | 359 | |||
XM_011534598.2 | XP_011532900.1 | 5 | 440 | |||
XM_017009654.1 | XP_016865143.1 | 5 | 588 | |||
XM_017009655.1 | XP_016865144.1 | 5 | 581 | |||
GenBank mRNA | AB033007.1 | - | 5 | 411 | ||
AF267855.1 | - | 3 | 367 | |||
AK026049.1 | - | 2 | 119 | |||
AK303453.1 | - | 2 | 277 | |||
AL136753.1 | - | 3 | 352 | |||
BC012766.2 | - | 5 | 348 | 3677787 | ||
BC014490.1 | - | 5 | 341 | 4907240 | ||
CR533474.1 | - | 3 | 137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
