siRNA Details
Gene Symbol: ERGIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031711.2 | NP_001026881.1 | 4 | 363 | ||
XM_011534597.1 | XP_011532899.1 | 4 | 256 | |||
XM_011534598.2 | XP_011532900.1 | 4 | 337 | |||
XM_017009654.1 | XP_016865143.1 | 4 | 485 | |||
XM_017009655.1 | XP_016865144.1 | 4 | 478 | |||
GenBank mRNA | AB033007.1 | - | 4 | 308 | ||
AF267855.1 | - | 2 | 264 | |||
AK026049.1 | - | 1 | 16 | |||
AK303453.1 | - | 1 | 25 | |||
AL136753.1 | - | 2 | 249 | |||
BC012766.2 | - | 4 | 245 | 3677787 | ||
BC014490.1 | - | 4 | 238 | 4907240 | ||
CR533474.1 | - | 2 | 34 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
