siRNA Details
Gene Symbol: ANKRD36B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025190.3 | NP_079466.3 | 39 | 3818 | ||
XM_005263995.4 | XP_005264052.1 | 39 | 4098 | |||
XM_006712661.1 | XP_006712724.1 | 39 | 4098 | |||
XM_017004578.1 | XP_016860067.1 | 39 | 4095 | |||
XM_017004579.1 | XP_016860068.1 | 37 | 3996 | |||
XM_017004580.1 | XP_016860069.1 | 37 | 3996 | |||
XM_017004581.1 | XP_016860070.1 | 37 | 3996 | |||
XM_017004582.1 | XP_016860071.1 | 37 | 3996 | |||
XM_017004583.1 | XP_016860072.1 | 38 | 3960 | |||
XM_017004584.1 | XP_016860073.1 | 33 | 3792 | |||
XM_017004585.1 | XP_016860074.1 | 37 | 3853 | |||
XM_017004586.1 | XP_016860075.1 | 34 | 2839 | |||
XM_017004587.1 | XP_016860076.1 | 38 | 4027 | |||
XM_017004588.1 | XP_016860077.1 | 37 | 3892 | |||
XM_017004589.1 | XP_016860078.1 | 36 | 3785 | |||
XM_017004590.1 | XP_016860079.1 | 35 | 3712 | |||
XM_017004591.1 | XP_016860080.1 | 21 | 3816 | |||
XM_017004592.1 | XP_016860081.1 | 40 | 4142 | |||
XM_017004593.1 | XP_016860082.1 | 38 | 3948 | |||
XM_017004594.1 | XP_016860083.1 | 37 | 3841 | |||
XM_017004595.1 | XP_016860084.1 | 38 | 3897 | |||
GenBank mRNA | AF432208.1 | - | 1 | 188 | ||
AF432209.1 | - | 1 | 290 | |||
AK056042.1 | - | 1 | 47 |
Gene Symbol: ANKRD26P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027248.3 | 14 | 2705 | |||
GenBank mRNA | BC036210.1 | - | 14 | 2705 |
Gene Symbol: ANKRD36BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026844.1 | 1 | 1142 | |||
GenBank mRNA | BC007072.1 | - | 1 | 1055 |
Gene Symbol: ANKRD36BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015424.1 | 13 | 1083 | |||
GenBank mRNA | AK097649.1 | - | 1 | 197 | ||
BC031698.1 | - | 13 | 1083 | |||
BX537549.1 | - | 2 | 95 | |||
BX648045.1 | - | 13 | 1117 |
Gene Symbol: ANKRD36C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310154.1 | NP_001297083.1 | 84 | 6523 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
