siRNA Details
Gene Symbol: RGPD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037866.1 | NP_001032955.1 | 6 | 1073 | ||
NM_001123363.3 | NP_001116835.1 | 6 | 1073 | |||
XM_011511766.2 | XP_011510068.1 | 5 | 657 | |||
XM_011511767.2 | XP_011510069.1 | 5 | 657 | |||
XM_011511769.2 | XP_011510071.1 | 5 | 657 | |||
XM_011511770.2 | XP_011510072.1 | 5 | 657 | |||
XM_017004839.1 | XP_016860328.1 | 5 | 657 | |||
XM_017004840.1 | XP_016860329.1 | 5 | 730 | |||
XM_017004841.1 | XP_016860330.1 | 5 | 860 | |||
XM_017004842.1 | XP_016860331.1 | 5 | 637 | |||
XM_017004843.1 | XP_016860332.1 | 5 | 657 | |||
XM_017004844.1 | XP_016860333.1 | 4 | 465 | |||
GenBank mRNA | AK056675.1 | - | 2 | 202 |
Gene Symbol: RANBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006267.4 | NP_006258.3 | 5 | 666 | ||
XM_005264002.2 | XP_005264059.1 | 5 | 701 | |||
XM_005264003.2 | XP_005264060.1 | 5 | 629 | |||
XM_005264004.2 | XP_005264061.1 | 5 | 629 | |||
XM_005264005.4 | XP_005264062.1 | 5 | 689 | |||
XM_005264007.2 | XP_005264064.1 | 5 | 629 | |||
XM_011511575.2 | XP_011509877.1 | 5 | 604 | |||
XM_011511576.2 | XP_011509878.1 | 5 | 629 | |||
XM_011511578.2 | XP_011509880.1 | 5 | 671 | |||
XM_017004623.1 | XP_016860112.1 | 5 | 629 | |||
XM_017004624.1 | XP_016860113.1 | 5 | 629 | |||
XM_017004625.1 | XP_016860114.1 | 5 | 701 | |||
GenBank mRNA | AB209483.1 | - | 2 | 282 | ||
D42063.1 | - | 5 | 667 |
Gene Symbol: RGPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144013.1 | NP_001137485.1 | 5 | 597 | ||
XM_017004738.1 | XP_016860227.1 | 6 | 646 | |||
XM_017004739.1 | XP_016860228.1 | 6 | 646 | |||
XM_017004740.1 | XP_016860229.1 | 5 | 571 |
Gene Symbol: RGPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182588.2 | NP_872394.2 | 5 | 622 | ||
XM_011511017.2 | XP_011509319.1 | 5 | 569 | |||
XM_011511018.2 | XP_011509320.1 | 5 | 572 | |||
XM_011511019.2 | XP_011509321.1 | 5 | 568 | |||
XM_011511020.2 | XP_011509322.1 | 5 | 568 | |||
XM_017003898.1 | XP_016859387.1 | 5 | 633 | |||
GenBank mRNA | BX537861.1 | - | 4 | 493 |
Gene Symbol: RGPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005054.2 | NP_005045.2 | 5 | 617 | ||
NM_032260.2 | NP_115636.1 | 6 | 1073 | |||
XM_006712794.3 | XP_006712857.2 | 5 | 648 | |||
XM_017005080.1 | XP_016860569.1 | 5 | 637 | |||
XM_017005081.1 | XP_016860570.1 | 5 | 648 | |||
XM_017005082.1 | XP_016860571.1 | 4 | 465 | |||
GenBank mRNA | AL136868.1 | - | 6 | 1074 | ||
EU794682.1 | - | 5 | 617 | |||
U64675.2 | - | 5 | 617 |
Gene Symbol: RGPD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164463.1 | NP_001157935.1 | 5 | 732 | ||
XM_005263747.4 | XP_005263804.1 | 5 | 730 | |||
XM_011511733.2 | XP_011510035.1 | 5 | 730 | |||
XM_017004825.1 | XP_016860314.1 | 5 | 730 | |||
XM_017004826.1 | XP_016860315.1 | 5 | 730 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
