siRNA Details
Gene Symbol: TRIM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033229.2 | NP_150232.2 | 1 | 645 | ||
XM_011514988.2 | XP_011513290.1 | 1 | 166 | |||
GenBank mRNA | AF220132.1 | - | 1 | 166 | ||
AF220133.1 | - | 1 | 166 | |||
AK290280.1 | - | 1 | 351 | |||
AK291853.1 | - | 1 | 303 | |||
BC038585.1 | - | 1 | 246 | 5758860 | ||
U34249.1 | - | 1 | 659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
