siRNA Details
Gene Symbol: TMEM236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098844.2 | NP_001092314.1 | 1 | 82 | ||
XM_011519626.2 | XP_011517928.1 | 1 | 233 | |||
GenBank mRNA | BC157856.1 | - | 1 | 82 | 9057075 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
