siRNA Details
Gene Symbol: CFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270420.1 | NP_001257349.1 | 1 | 203 | ||
NM_001270421.1 | NP_001257350.1 | 1 | 203 | |||
NM_032545.3 | NP_115934.1 | 1 | 203 | |||
XM_011511486.2 | XP_011509788.1 | 1 | 198 | |||
GenBank mRNA | AK315326.1 | - | 1 | 203 | ||
BC110080.1 | - | 1 | 76 | 40030641 | ||
BC146897.1 | - | 1 | 80 | 9056825 | ||
BM509218.1 | - | 1 | 137 | |||
CA777249.1 | - | 1 | 176 | |||
CA941621.1 | - | 1 | 135 |
Gene Symbol: CFC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079530.1 | NP_001072998.1 | 1 | 306 | ||
XM_005263737.4 | XP_005263794.1 | 1 | 188 | |||
GenBank mRNA | BQ777283.1 | - | 1 | 148 | ||
BU784123.1 | - | 1 | 161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
