siRNA Details
Gene Symbol: JPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271604.2 | NP_001258533.1 | 1 | 438 | ||
NM_001271605.1 | NP_001258534.1 | 1 | 438 | |||
NM_020655.3 | NP_065706.2 | 1 | 438 | |||
XM_017023483.1 | XP_016878972.1 | 1 | 464 | |||
XM_017023484.1 | XP_016878973.1 | 1 | 464 | |||
GenBank mRNA | AB042636.1 | - | 1 | 347 | ||
AB593088.1 | - | 1 | 438 | |||
BC036533.2 | - | 1 | 420 | 5264853 | ||
BI553267.1 | - | 1 | 437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
