siRNA Details
Gene Symbol: ZNF395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018660.2 | NP_061130.1 | 10 | 2991 | ||
GenBank mRNA | AK026076.1 | - | 1 | 858 | ||
AK312078.1 | - | 1 | 381 | |||
AL137588.1 | - | 1 | 307 | |||
BC002919.1 | - | 1 | 543 | |||
BC008870.2 | - | 1 | 257 | |||
BC063660.1 | - | 1 | 162 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
