siRNA Details
Gene Symbol: KRT23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015515.4 | NP_056330.3 | 2 | 899 | ||
GenBank mRNA | AF102848.1 | - | 1 | 327 | ||
AK002047.1 | - | 1 | 311 | |||
AK225866.1 | - | 1 | 682 | |||
AK289449.1 | - | 1 | 394 | |||
AK310880.1 | - | 1 | 483 | |||
AK314604.1 | - | 1 | 111 | |||
DA846104.1 | - | 1 | 111 |
Gene Symbol: KRT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000526.4 | NP_000517.2 | 1 | 441 | ||
GenBank mRNA | AK309280.1 | - | 1 | 441 | ||
BC002690.2 | - | 1 | 426 | 3610489 | ||
BC019097.2 | - | 1 | 432 | 5013623 | ||
BC042437.1 | - | 1 | 396 | 4776763 | ||
BC094830.1 | - | 1 | 432 | 5443153 | ||
BT007186.1 | - | 1 | 380 | |||
DB018715.1 | - | 1 | 441 |
Gene Symbol: KRT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002275.3 | NP_002266.2 | 1 | 541 | ||
XM_011524784.2 | XP_011523086.1 | 2 | 2124 | |||
XM_017024614.1 | XP_016880103.1 | 1 | 587 | |||
GenBank mRNA | AK090604.1 | - | 3 | 1094 | ||
AK122864.1 | - | 1 | 3936 | |||
AK304059.1 | - | 1 | 413 | |||
AL832226.1 | - | 1 | 2542 | |||
BC002641.2 | - | 1 | 406 | 3610392 | ||
BT007261.1 | - | 1 | 350 | |||
X07696.1 | - | 1 | 411 |
Gene Symbol: KRT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005557.3 | NP_005548.2 | 1 | 525 | ||
GenBank mRNA | AF061812.1 | - | 1 | 400 | ||
AK290853.1 | - | 1 | 463 | |||
AK309278.1 | - | 1 | 465 | |||
AK309296.1 | - | 1 | 463 | |||
BC039169.1 | - | 1 | 436 | 4752995 |
Gene Symbol: KRT16P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK301679.1 | - | 1 | 465 |
Gene Symbol: KRT16P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029392.1 | 1 | 468 | |||
GenBank mRNA | AK304304.1 | - | 1 | 468 |
Gene Symbol: KRT16P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029393.1 | 1 | 463 | |||
GenBank mRNA | BC110641.1 | - | 1 | 446 | ||
DB013784.1 | - | 1 | 495 |
Gene Symbol: KRT17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000422.2 | NP_000413.1 | 1 | 408 | ||
GenBank mRNA | AK095342.1 | - | 1 | 337 | ||
AK304373.1 | - | 1 | 355 | |||
BC000159.2 | - | 1 | 337 | 2986100 | ||
BC011901.2 | - | 1 | 335 | 4111703 | ||
BC056421.1 | - | 1 | 337 | 5208304 | ||
BC072019.1 | - | 1 | 333 | 5442690 | ||
BP226061.1 | - | 1 | 408 | |||
BX647923.1 | - | 1 | 74 | |||
X62571.1 | - | 1 | 335 |
Gene Symbol: KRT17P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK296148.1 | - | 1 | 356 |
Gene Symbol: KRT19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002276.4 | NP_002267.2 | 1 | 417 | ||
GenBank mRNA | AK313261.1 | - | 1 | 336 | ||
BC010409.2 | - | 1 | 328 | 3689178 | ||
BC067744.1 | - | 1 | 336 | 30332499 | ||
BC084574.1 | - | 1 | 319 | 6730698 | ||
BG490639.1 | - | 1 | 321 | |||
BG750971.1 | - | 410 | ||||
BP328445.1 | - | 1 | 424 | |||
DA068330.1 | - | 1 | 400 | |||
Y00503.1 | - | 1 | 307 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133730, NM_010663, NM_008469, NM_010666, AB013607, and others.
Gene Ontology Categories:
