siRNA Details
Gene Symbol: TSPY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022573.2 | NP_072095.2 | 4 | 830 | ||
XM_005262569.4 | XP_005262626.1 | 4 | 790 | |||
XM_017030062.1 | XP_016885551.1 | 4 | 783 | |||
GenBank mRNA | AK307425.1 | - | 4 | 735 |
Gene Symbol: TSPY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197242.2 | NP_001184171.1 | 4 | 830 | ||
NM_001320964.1 | NP_001307893.1 | 5 | 569 | |||
NM_003308.4 | NP_003299.2 | 4 | 830 | |||
XM_017030064.1 | XP_016885553.1 | 4 | 798 | |||
GenBank mRNA | BC121113.1 | - | 5 | 573 | 40120433 | |
M98525.1 | - | 4 | 734 | |||
U58096.1 | - | 4 | 829 |
Gene Symbol: TSPY10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282469.2 | NP_001269398.1 | 4 | 830 | ||
NM_001320962.1 | NP_001307891.1 | 4 | 830 | |||
XM_017030025.1 | XP_016885514.1 | 4 | 798 | |||
GenBank mRNA | AK308271.1 | - | 3 | 679 | ||
AY130858.1 | - | 4 | 784 | |||
BC075016.2 | - | 4 | 811 | 30915406 | ||
BC121114.1 | - | 4 | 834 | 40120435 | ||
DB545400.1 | - | 2 | 164 |
Gene Symbol: TSPY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077697.2 | NP_001071165.2 | 4 | 830 | ||
XM_005272742.4 | XP_005272799.1 | 4 | 805 | |||
XM_011544004.2 | XP_011542306.1 | 4 | 798 |
Gene Symbol: TSPY4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164471.1 | NP_001157943.1 | 4 | 802 | ||
XM_005272743.4 | XP_005272800.1 | 4 | 823 | |||
XM_017030065.1 | XP_016885554.1 | 4 | 816 | |||
GenBank mRNA | AK093413.1 | - | 3 | 903 | ||
BX281192.1 | - | 3 | 296 |
Gene Symbol: TSPY8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243721.1 | NP_001230650.1 | 4 | 784 | ||
XM_005272744.4 | XP_005272801.1 | 4 | 798 | |||
XM_011544005.2 | XP_011542307.1 | 4 | 791 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
