siRNA Details
Gene Symbol: CLEC18C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173619.3 | NP_775890.2 | 5 | 737 | ||
XM_006721179.1 | XP_006721242.1 | 5 | 833 | |||
XM_011523034.1 | XP_011521336.1 | 6 | 1079 | |||
XM_011523035.1 | XP_011521337.1 | 5 | 833 | |||
XM_011523036.2 | XP_011521338.1 | 4 | 663 | |||
XM_011523037.1 | XP_011521339.1 | 5 | 833 | |||
GenBank mRNA | AL833339.1 | - | 5 | 824 | ||
BC039068.1 | - | 5 | 885 | 5188643 |
Gene Symbol: CLEC18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136214.2 | NP_001129686.1 | 6 | 769 | ||
NM_001271197.1 | NP_001258126.1 | 6 | 849 | |||
NM_182619.3 | NP_872425.2 | 6 | 875 | |||
XM_005255934.1 | XP_005255991.1 | 5 | 818 | |||
XM_017023206.1 | XP_016878695.1 | 5 | 818 | |||
XM_017023207.1 | XP_016878696.1 | 4 | 648 | |||
XM_017023208.1 | XP_016878697.1 | 5 | 1001 | |||
XM_017023209.1 | XP_016878698.1 | 5 | 818 | |||
GenBank mRNA | AF428259.1 | - | 5 | 888 | ||
AF521893.1 | - | 6 | 888 | |||
AK074773.1 | - | 5 | 703 | |||
AK289884.1 | - | 5 | 817 | |||
AK298310.1 | - | 5 | 721 | |||
BC078143.1 | - | 6 | 751 | 5761001 | ||
BC141808.1 | - | 6 | 849 | 40147524 |
Gene Symbol: CLEC18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011880.2 | NP_001011880.2 | 5 | 831 | ||
XM_005255976.2 | XP_005256033.1 | 5 | 908 | |||
XM_011523093.2 | XP_011521395.1 | 4 | 614 | |||
XM_017023255.1 | XP_016878744.1 | 6 | 1085 | |||
XM_017023256.1 | XP_016878745.1 | 5 | 908 | |||
GenBank mRNA | AK293982.1 | - | 4 | 591 | ||
AY358373.1 | - | 5 | 753 | |||
BM695392.1 | - | 1 | 42 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
