siRNA Details
Gene Symbol: CLEC18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136214.2 | NP_001129686.1 | 13 | 1599 | ||
NM_001271197.1 | NP_001258126.1 | 13 | 1679 | |||
NM_182619.3 | NP_872425.2 | 13 | 1705 | |||
XM_005255934.1 | XP_005255991.1 | 12 | 1648 | |||
GenBank mRNA | AF428259.1 | - | 12 | 1716 | ||
AF521893.1 | - | 13 | 1718 | |||
AK074773.1 | - | 12 | 1533 | |||
AK289884.1 | - | 12 | 1647 | |||
BC008616.1 | - | 3 | 593 | |||
BC078143.1 | - | 13 | 1581 | 5761001 | ||
BC141808.1 | - | 13 | 1679 | 40147524 |
Gene Symbol: CLEC18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011880.2 | NP_001011880.2 | 13 | 1688 | ||
XM_005255976.2 | XP_005256033.1 | 12 | 1738 | |||
XM_011523093.2 | XP_011521395.1 | 11 | 1444 | |||
XM_017023255.1 | XP_016878744.1 | 13 | 1915 | |||
GenBank mRNA | AY358373.1 | - | 13 | 1610 |
Gene Symbol: CLEC18C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173619.3 | NP_775890.2 | 12 | 1567 | ||
XM_006721179.1 | XP_006721242.1 | 12 | 1663 | |||
XM_011523034.1 | XP_011521336.1 | 13 | 1909 | |||
XM_011523036.2 | XP_011521338.1 | 11 | 1493 | |||
GenBank mRNA | AF448856.1 | - | 12 | 1714 | ||
AL833339.1 | - | 9 | 2937 | |||
BC039068.1 | - | 10 | 2621 | 5188643 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
