siRNA Details
Gene Symbol: DMRTC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080851.1 | NP_001074320.1 | 1 | 94 | ||
XM_006724684.2 | XP_006724747.1 | 2 | 173 | |||
XM_006724685.3 | XP_006724748.1 | 2 | 198 | |||
XM_017029776.1 | XP_016885265.1 | 2 | 204 |
Gene Symbol: DMRTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033053.2 | NP_149042.2 | 1 | 94 | ||
XM_005262287.3 | XP_005262344.1 | 2 | 173 | |||
XM_005262288.3 | XP_005262345.1 | 2 | 198 | |||
XM_017029725.1 | XP_016885214.1 | 2 | 204 | |||
GenBank mRNA | AJ291670.1 | - | 1 | 280 | ||
BC029799.1 | - | 2 | 334 | |||
BC047596.1 | - | 2 | 237 | 4830694 | ||
CB121638.1 | - | 2 | 195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
