siRNA Details
Gene Symbol: HBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000518.4 | NP_000509.1 | 1 | 37 | ||
GenBank mRNA | AF117710.1 | - | 1 | 17 | ||
AF181989.1 | - | 1 | 20 | |||
AF349114.1 | - | 1 | 22 | |||
AK311825.1 | - | 1 | 37 | |||
AY509193.1 | - | 1 | 37 | |||
BC007075.1 | - | 1 | 38 | 4292125 | ||
EU694432.1 | - | 1 | 37 | |||
M14574.1 | - | 1 | 49 | |||
V00497.1 | - | 1 | 37 |
Gene Symbol: HBD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000519.3 | NP_000510.1 | 1 | 182 | ||
GenBank mRNA | BC069307.1 | - | 1 | 37 | 7262103 | |
BC070282.1 | - | 1 | 38 | 30418964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
