siRNA Details
Gene Symbol: HBD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000519.3 | NP_000510.1 | 1 | 205 | ||
GenBank mRNA | AY034468.1 | - | 1 | 205 | ||
BC069307.1 | - | 1 | 60 | 7262103 | ||
BC070282.1 | - | 1 | 61 | 30418964 | ||
BU664913.1 | - | 1 | 24 |
Gene Symbol: HBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000518.4 | NP_000509.1 | 1 | 60 | ||
GenBank mRNA | AF181989.1 | - | 1 | 43 | ||
AK311825.1 | - | 1 | 60 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
