siRNA Details
Gene Symbol: RHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB049754.1 | - | 2 | 172 | ||
AF177938.1 | - | 2 | 172 | |||
AF187846.1 | - | 2 | 172 | |||
FR745438.1 | - | 2 | 172 | |||
JF436967.1 | - | 2 | 172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
