siRNA Details
Gene Symbol: RPL21P28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026911.1 | 1 | 362 | |||
GenBank mRNA | EC535637.1 | - | 1 | 29 |
Gene Symbol: RPL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000982.3 | NP_000973.2 | 1 | 360 | ||
GenBank mRNA | AF086172.1 | - | 1 | 259 | ||
AK130968.1 | - | 1 | 359 | |||
AK311795.1 | - | 1 | 357 | |||
AY320407.1 | - | 1 | 360 | |||
BC001603.1 | - | 1 | 342 | 3543702 | ||
BC007505.2 | - | 1 | 323 | 2964441 | ||
BC019002.2 | - | 1 | 315 | |||
BC062981.1 | - | 1 | 340 | 6084268 | ||
BC070184.1 | - | 1 | 344 | 4281405 | ||
BC070323.1 | - | 1 | 357 | 4248655 | ||
BC070330.1 | - | 1 | 357 | 3931616 | ||
BC071902.1 | - | 1 | 340 | 4901492 | ||
BC104478.1 | - | 2 | 285 | |||
BC104667.1 | - | 1 | 361 | 6724053 | ||
BX647669.1 | - | 1 | 359 | |||
CR457032.1 | - | 1 | 317 | |||
U14967.1 | - | 1 | 346 | |||
X89401.1 | - | 1 | 330 |
Gene Symbol: RPL21P19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209100.1 | - | 1 | 419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
