siRNA Details
Gene Symbol: RASEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152573.3 | NP_689786.2 | 1 | 140 | ||
XM_005251730.3 | XP_005251787.1 | 1 | 189 | |||
GenBank mRNA | AK056176.1 | - | 1 | 190 | ||
BC023566.1 | - | 1 | 140 | 4138119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
