siRNA Details
Gene Symbol: RPL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000982.3 | NP_000973.2 | 1 | 336 | ||
GenBank mRNA | AF086172.1 | - | 1 | 235 | ||
AK130968.1 | - | 1 | 335 | |||
AK311795.1 | - | 1 | 333 | |||
BC001603.1 | - | 1 | 318 | 3543702 | ||
BC007505.2 | - | 1 | 299 | 2964441 | ||
BC019002.2 | - | 1 | 291 | |||
BC062981.1 | - | 1 | 316 | 6084268 | ||
BC070184.1 | - | 1 | 320 | 4281405 | ||
BC070185.1 | - | 1 | 333 | |||
BC070323.1 | - | 1 | 333 | 4248655 | ||
BC070330.1 | - | 1 | 333 | 3931616 | ||
BC071902.1 | - | 1 | 316 | 4901492 | ||
BC104478.1 | - | 2 | 261 | |||
BC104667.1 | - | 1 | 337 | 6724053 | ||
BX647669.1 | - | 1 | 335 | |||
CR457032.1 | - | 1 | 293 | |||
U25789.1 | - | 1 | 326 | |||
X89401.1 | - | 1 | 306 |
Gene Symbol: RPL21P19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209100.1 | - | 1 | 395 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
