siRNA Details
Gene Symbol: GOLGA6L5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003246.4 | 6 | 667 | |||
GenBank mRNA | AK092490.1 | - | 3 | 280 | ||
AK097432.1 | - | 6 | 669 | |||
AK130562.1 | - | 5 | 755 | |||
DA284832.1 | - | 5 | 455 |
Gene Symbol: GOLGA6L10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164465.3 | NP_001157937.2 | 6 | 705 | ||
NM_001322400.1 | NP_001309329.1 | 6 | 705 | |||
GenBank mRNA | AK302092.1 | - | 6 | 751 | ||
CR936602.1 | - | 5 | 1332 |
Gene Symbol: GOLGA6L17P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111962.1 | 6 | 669 | |||
GenBank mRNA | AK301885.1 | - | 669 |
Gene Symbol: GOLGA6L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310153.1 | NP_001297082.1 | 6 | 667 | ||
GenBank mRNA | AK301968.1 | - | 6 | 634 |
Gene Symbol: GOLGA6L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267536.2 | NP_001254465.2 | 6 | 667 | ||
XM_017022481.1 | XP_016877970.1 | 6 | 667 | |||
XM_017022482.1 | XP_016877971.1 | 6 | 667 |
Gene Symbol: GOLGA6L9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291420.1 | NP_001278349.1 | 5 | 573 | ||
NM_198181.3 | NP_937824.3 | 6 | 705 | |||
XM_011521601.2 | XP_011519903.1 | 5 | 1256 | |||
GenBank mRNA | AK128268.1 | - | 2 | 254 | ||
BC041854.1 | - | 5 | 573 | 5270411 | ||
BY996849.1 | - | 3 | 448 | |||
CR936812.1 | - | 3 | 3109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
