siRNA Details
Gene Symbol: ARL6IP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002251.2 | NP_001002251.2 | 2 | 783 | ||
NM_001002252.2 | NP_001002252.2 | 2 | 783 | |||
NM_001278378.1 | NP_001265307.1 | 2 | 783 | |||
NM_001278379.1 | NP_001265308.1 | 2 | 423 | |||
NM_001278380.1 | NP_001265309.1 | 2 | 925 | |||
NM_016638.3 | NP_057722.3 | 2 | 783 | |||
NM_018694.3 | NP_061164.3 | 2 | 783 | |||
NR_103512.1 | 2 | 783 | ||||
GenBank mRNA | AB035384.1 | - | 2 | 148 | ||
AF267748.1 | - | 2 | 259 | |||
AK027285.1 | - | 2 | 157 | |||
AK315759.1 | - | 2 | 139 | |||
BC001958.1 | - | 2 | 117 | 3544177 | ||
BC015569.2 | - | 2 | 113 | 3609127 | ||
BC015909.1 | - | 2 | 783 | 4648478 | ||
BC094839.1 | - | 2 | 517 | 6339167 | ||
BQ643520.1 | - | 2 | 220 | |||
CD672894.1 | - | 2 | 133 | |||
DQ099385.1 | - | 1 | 86 | |||
EF036485.1 | - | 1 | 71 | |||
EU624490.1 | - | 2 | 783 | |||
EU624491.1 | - | 2 | 490 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
