siRNA Details
Gene Symbol: SPATA33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271907.1 | NP_001258836.1 | 2 | 230 | ||
NM_001271908.1 | NP_001258837.1 | 2 | 148 | |||
NM_001271909.1 | NP_001258838.1 | 1 | 286 | |||
NM_001271910.1 | NP_001258839.1 | 2 | 230 | |||
NM_153025.2 | NP_694570.1 | 2 | 177 | |||
NR_073516.1 | 1 | 286 | ||||
XM_006721133.2 | XP_006721196.1 | 2 | 230 | |||
GenBank mRNA | AK056168.1 | - | 2 | 119 | ||
AK303024.1 | - | 2 | 196 | |||
BC045670.1 | - | 2 | 163 | |||
BC148299.2 | - | 2 | 119 | 40112476 | ||
BP345094.1 | - | 1 | 286 | |||
DA300380.1 | - | 2 | 140 | |||
DB025192.1 | - | 2 | 177 | |||
DB452323.1 | - | 2 | 230 | |||
DC357681.1 | - | 2 | 188 | |||
DC374690.1 | - | 2 | 198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
