siRNA Details
Gene Symbol: CGB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033043.1 | NP_149032.1 | 1 | 83 | ||
GenBank mRNA | BC006290.2 | - | 1 | 44 | 3960125 | |
BC051378.1 | - | 1 | 44 | 3956670 |
Gene Symbol: CGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000737.3 | NP_000728.1 | 1 | 122 | ||
GenBank mRNA | AK291552.1 | - | 1 | 80 | ||
BC022796.1 | - | 1 | 60 | 4811294 | ||
BC106059.1 | - | 1 | 68 | 4992804 | ||
BX334905.2 | - | 1 | 122 |
Gene Symbol: CGB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092722.1 | - | 3 | 1153 |
Gene Symbol: CGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033183.2 | NP_149439.1 | 1 | 90 | ||
GenBank mRNA | BC030994.1 | - | 1 | 85 | 4605021 | |
BX118262.1 | - | 1 | 90 | |||
BX380841.2 | - | 1 | 72 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
